Learn More
Astell Scientific™ Integral Steam Generator
Improve cycle times by increasing the efficiency of steam production. Astell Scientific™ Integral Steam Generator system also includes automatic water filling.
Brand:  Astell Scientific™ 24KWSG
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15791439
  • £5430.00 / Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
Description
Description
  • Removes the heating elements from the chamber and replaces them with an integral generator for improved control over steam production.
  • Autofill is also fitted as standard to the steam generator; eliminating the need to manually refill the chamber between cycles.
  • A mains water supply enters the steam generator, where it is heated and converted to steam. This then enters the chamber directly from the steam generator as and when it is required.
Specifications
Specifications
Autoclaves of 120 L Capacity and Above (Excluding The E-Range).
Integral Steam Generator
Automatic water filling
24 kw