Learn More
Integrin Beta 3 Mouse anti-Human, Unconjugated, Clone: 3D8E5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66952-1-IG-150UL
This item is not returnable.
View return policy
Description
The Integrin Beta 3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Integrin Beta 3. This antibody recognizes human antigen. The Integrin Beta 3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Integrin Beta 3 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry | |
3D8E5 | |
Unconjugated | |
150 μL | |
3690 | |
IgG2b |