Learn More
Internal Standard, For USEPA Method 525.2, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep 5252-I
Code : NEW
Additional Details : Weight : 0.00100kg Transport : UN number : 1090 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Acenaphthene-d10 | 15067-26-2 | 500μg/mL |
Chrysene-d12 | 1719-03-5 | 500μg/mL |
Phenanthrene-d10 | 1517-22-2 | 500μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Internal Standard | |
2mL amber prescored glass ampule | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
For US EPA Method 525 |
USEPA Method 525.2 | |
1 mL | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation | |
Acetone |
Safety and Handling
- Internal Standard
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetone
Product intended for laboratory use only