missing translation for 'onlineSavingsMsg'
Learn More

Internal Standard, 4-Bromo-3,5-dimethylphenyl-n-methylcarbamate, SPEX CertiPrep™

Product Code. 15265796
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
4-Bromo-3,5-dimethylphenyl-N-methylcarbamate672-99-1100μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 672-99-1, 75-05-8
CAS Min % 0.01, 99.99
CAS Max % 0.01, 99.99
Molecular Formula C10H12BrNO2
MDL Number MFCD00027941
InChI Key ZOZFMTULOYRWEL-UHFFFAOYSA-N
SMILES CNC(=O)OC1=CC(C)=C(Br)C(C)=C1
IUPAC Name (4-bromo-3,5-dimethylphenyl) N-methylcarbamate
Molecular Weight (g/mol) 258.12
PubChem CID 688148
Packaging 2mL amber prescored glass ampule
Methods For US EPA Methods 531, 531.1
Solvent or Matrix Acetonitrile
Quantity 1 mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material 4-Bromo-3,5-dimethylphenyl-n-methylcarbamate, USEPA Method 531.1
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Internal Standard
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'