Learn More
STEMCELL Technologies IntestiCult™ Organoid Growth Medium (Human)
Cell culture medium for establishment and maintenance of human intestinal organoids
Brand: STEMCELL Technologies 06010
Additional Details : Weight : 0.24440kg
Description
- Model key characteristics of the adult intestinal epithelium using intestinal organoids established and maintained with this complete medium formulation and optimized protocol
- Using the easy-to-follow and robust protocol, you can derive organoids from human intestinal crypts in one week; organoid growth across donor samples, including those that are otherwise difficult to grow, is enabled by an enriched stem cell population
- Organoids grown in IntestiCult Organoid Growth Medium (Human) incorporate a functional lumen enclosed by a polarized intestinal epithelial cell layer and, for versatile modeling applications, can be further differentiated in 3D or in 2D as submerged monolayers or air-liquid interface (ALI) cultures using IntestiCult Organoid Differentiation Medium (Human; Catalog #100-0214)
Specifications
Intestinal Cells | |
Cell Culture; Differentiation; Expansion; Maintenance; Organoid Culture | |
1 Kit |