Learn More
STEMCELL Technologies IntestiCult™ Organoid Growth Medium (Mouse)
Cell culture medium for establishment and maintenance of mouse intestinal organoids
Brand: STEMCELL Technologies 6005
Additional Details : Weight : 0.00001kg
Description
- IntestiCult™ Organoid Growth Medium (Mouse) is a defined, serum-free cell culture medium for efficient establishment and long-term maintenance of mouse intestinal organoids
- These organoids, or 'mini-guts', provide a convenient in vitro organotypic culture system for studying both the small and large intestinal epithelium and associated stem cell dynamics
- Organoids grown in IntestiCult™ feature a polarized epithelium that contains all of the known cell types of the adult intestinal epithelium
- Individual intestinal crypts rapidly form organoids when cultured in IntestiCult™ Organoid Growth Medium (Mouse)
- Applications of these cultures include studying the development and function of the normal and tumorigenic intestinal epithelium, modeling intestinal disease, and investigating stem cell properties and regenerative therapy approaches
- Organoid culture enables convenient in vitro characterization of a system with strong physiological relevance to the adult intestine
Specifications
Intestinal Cells | |
Cell Culture; Differentiation; Expansion; Maintenance; Organoid Culture | |
1 Kit |