missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ intestinal differentiation media pack, kit

Brand:  Falcon™ 355058

Code : 67

Additional Details : Weight : 1.48000kg

Product Code. 10506771

  • £276.02 / 1 set
Estimated Shipment: 26-11-2024
to see stock.

Description

Description

Corning Intestinal Differentiation Media Pack contains Corning Seeding Basal Medium (500mL), Differentiation Medium (500mL), and Corning MITO+ Serum Extender (2 x 0.5mL), all in one convenient kit.
Specifications

Specifications

1 Pack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers