Learn More
Invitrogen™ eBioscience™ Intracellular Fixation & Permeabilization Buffer Set
Designed for use in intracellular staining and flow cytometric analysis and has been specially formulated to reduce non-specific staining of fluorochrome-labelled antibodies and increase fluorescence signal-to-noise ratios.
Brand: Invitrogen™ 88-8824-00
96.48 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Alert:
Contains Sodium azide and formaldehyde
Includes:
- IC Fixation Buffer
- Permeabilization Buffer (10X)
Description
- In the first step, live cells can be 'fixed' with the Fixation Buffer, which cross- links proteins
- The second step, using the Permeabilization Buffer, creates holes in the membrane and allows the intracellular staining antibodies to enter the cell effectively
- Subsequent washing steps, antibody additions, and incubations after cell permeabilization should be performed using the Permeabilization Buffer
- Final washing step and cell resuspension prior to running the sample is done using Flow Cytometry Staining Buffer (cat. 00-4222).
- This kit is commonly used to stain cytoplasmic proteins and proteins in the secretory pathway such as chemokines and cytokines
- Not recommended for intranuclear staining. Instead, please refer to the Foxp3/Transcription Factor Buffer Set for nuclear staining
- Before the fixation step, it is recommended to label the live cells using one of the Fixable Viability Dyes and then proceed with fixation

Specifications
eBioscience™ | |
Fixn/Perm Buffer Set | |
1 kit | |
Fixation/Permeabilization Buffer |
Safety and Handling
- eBioscience Intracellular Fixation & Permeabilization Buffer Set
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Paraformaldehyde
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2990891 | Certificate of Analysis | 31/01/2025 | 88-8824-00 |
2504455 | Certificate of Analysis | 29/01/2025 | 88-8824-00 |
2528101 | Certificate of Analysis | 29/01/2025 | 88-8824-00 |
2555740 | Certificate of Analysis | 29/01/2025 | 88-8824-00 |
2589190 | Certificate of Analysis | 29/01/2025 | 88-8824-00 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.