Learn More
INTS3 Mouse anti-Human, Unconjugated, Clone: 2G11B7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67060-1-IG-150UL
 View more versions of this product
Product Code. 16819465
  • £349.00 / 150µL
Explore more special offers
Description
Description
The INTS3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human INTS3. This antibody recognizes human, mouse, rat antigen. The INTS3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
INTS3
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence, ELISA
2G11B7
Unconjugated
150 μL
65123
IgG2b