missing translation for 'onlineSavingsMsg'
Learn More

Inulin From Chicory Root MP Biomedicals

£1510.00

Chemical Identifiers

CAS 9005-80-5
Molecular Formula (C6H10O5)n
Synonym Dahlin, Alantin
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11466921
View Documents
MP Biomedicals
0219897191
1 kg
£1510.00
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

9005-80-5
(C6H10O5)n
Dahlin, Alantin

Specifications

9005-80-5
(C6H10O5)n
Dahlin, Alantin
Inulin, From Chicory Root
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.