Learn More
Invar∣r, Iron Nickel powder, -325 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
£46.10 - £143.00
Specifications
Chemical Name or Material | Invar™ Iron Nickel powder |
---|---|
Color | Gray |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Ni 65:35 wt% |
Assay Percent Notes | (metals basis) |
MDL Number | MFCD01741928 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11405390
|
Thermo Scientific Alfa Aesar
044600.22 |
100 g | |||||||
11415390
|
Thermo Scientific Alfa Aesar
044600.36 |
500 g |
£143.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Invar is used where high dimensional stability is required, such as precision instruments, clocks, seismic creep gauges, television shadow-mask frames, valves in motors, and antimagnetic watches.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Invar™ Iron Nickel powder | |
Fe:Ni 65:35 wt% | |
MFCD01741928 | |
99.9% | |
Odorless |
Gray | |
(metals basis) | |
UN3089 | |
−325 Mesh | |
Powder |
Safety and Handling
GHS H Statement
H228-H372-H351-H319-H317-H335
Flammable solid.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
P210-P260-P261-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
missing translation for 'tsca' : TSCA
- Invar, Iron Nickel powder
Signal Word
- Danger
Hazard Category
- Flammable solid Category 1
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only