missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 1 Kb Plus DNA Ladder
GREENER_CHOICE

Powerful tool for estimating molecular weight of linear, double-stranded DNA fragments

Brand:  Invitrogen™ 10787026

490.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11573127

  • £700.00 / 1000µg
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Requires : Gel-loading buffer (Invitrogen part no. 10816015)
This item is not returnable. View return policy

Description

Description

One Kb Plus DNA ladder is composed of 20 highly purified, double-stranded DNA bands spanning 100bp to 12,000bp. This ladder has 12 evenly spaced bands ranging from 1Kb to 12Kb, quick orientation band at 1,650bp that forms distinct doublet with 2Kb band, and seven bands of round sizes below 1Kb. One Kb Plus DNA ladder is compatible with agarose gels cast in lab as well as E-Gel™ pre-cast agarose gels. One Kb Plus DNA ladder is most popular DNA ladder used to estimate molecular mass of double-stranded DNA bands from PCR, restriction digests, and other molecular biology protocols. Unlike some other DNA ladders, 1Kb Plus DNA ladder has been specially designed with bands of round, easy-to-remember sizes. Logical band layout, and quick identification doublet, allow to estimate size of band at glance.

  • Round, easy-to-remember band sizes
  • Broad size range (100bp–12Kb)
  • Compatible with all agarose gels
  • Less waste, sustainable packaging

  • Popular and easiest-to-use ladder
  • Single ladder for most of needs
  • Simplified format for use with all DNA agarose gels
  • DNA stain and radiolabel compatible

Acrylamide Gel Electrophoresis, Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 4 x 500 µL 1 Kb Plus DNA Ladder
• 4 x 1 mL 10X BlueJuice Gel Loading Buffer

Store at -20°C.

Agarose
No
100 to 15000 bp
Agarose gel
2000 Applications
Approved for shipment at Room Temperature or on Dry Ice
0.5 μg/μL
DNA Ladder
1 mL
500 μL
Sustainable packaging
4 x 500 μL DNA Ladder, 4 x 1 mL Sample Loading Buffer, 2000 Applications
Individual chromatography-purified DNA fragments
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.