Learn More
Invitrogen™ 100 bp DNA Ladder
Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp.
Brand: Invitrogen™ 15628019
73.64 GBP valid until 2025-02-28
Use promo code "24154" to get your promotional price.
Description
Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. 100 bp DNA Ladder consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 1500, and 600 bp for easy orientation.
100 bp DNA Ladder is ideal for separation on 1–2% agarose gels.
Highlights of 100 bp DNA Ladder:
• Sharp, clear bands—chromatography purified fragments for consistent and reliable results
• Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration
• Precise—an exact amount of DNA in each band
Product use
The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.
This ladder can be radiolabeled with T4 polynucleotide kinase or T4 DNA polymerase.
Order Info
Shipping Condition: Room Temperature
Specifications
• 100 µL 100 bp DNA Ladder • 1 mL 10X BlueJuice Gel Loading Buffer Store at -20°C. |
|
Agarose | |
No | |
100 to 2000 bp | |
Agarose gel | |
100 Applications | |
Approved for shipment at Room Temperature or on Dry Ice |
0.5 μg/μL | |
DNA Ladder | |
1 mL | |
100 μL | |
Sustainable packaging | |
100 μL DNA Ladder, 1 mL 10X Sample Loading Buffer, 100 Applications | |
Individual chromatography-purified DNA fragments |
Special Offers
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24154Expires: 2025-02-28
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.