missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 100 bp DNA Ladder
GREENER_CHOICE

Product Code. p-4924091
Click to view available options
Quantity:
100 Applications
500 Applications
Unit Size:
250µg
50µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11538766

Brand: Invitrogen™ 15628019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp.

Invitrogen 100 bp DNA Ladder is designed for sizing and approximate quantification of double-stranded DNA in the range of 100 bp to 2,000 bp. 100 bp DNA Ladder consists of 13 individual chromatography-purified DNA fragments and has reference bands at 2000, 1500, and 600 bp for easy orientation.

100 bp DNA Ladder is ideal for separation on 1–2% agarose gels.

Highlights of 100 bp DNA Ladder:
Sharp, clear bands—chromatography purified fragments for consistent and reliable results
Convenient—provided with 10X BlueJuice Gel Loading Buffer for tracking of sample DNA migration
Precise—an exact amount of DNA in each band

Product use
The double-stranded DNA ladder can be visualized on 1–2% agarose gels after ethidium bromide or SYBR Safe staining. The ladder is designed with a uniform intensity of DNA bands for a clear view of each band. An exact amount of DNA in each band allows approximate quantification of DNA samples.

This ladder can be radiolabeled with T4 polynucleotide kinase or T4 DNA polymerase.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 100 µL 100 bp DNA Ladder
• 1 mL 10X BlueJuice Gel Loading Buffer

Store at -20°C.

Concentration 0.5 μg/μL
Gel Type Agarose
Product Type DNA Ladder
Ready to Load No
Sample Loading Volume 1 mL
Size Range 100 to 2000 bp
Volume (Metric) 100 μL
Gel Compatibility Agarose gel
Green Features Sustainable packaging
Kit Contents 100 μL DNA Ladder, 1 mL 10X Sample Loading Buffer, 100 Applications
No. of Reactions 100 Applications
Quantity 100 Applications
Shipping Condition Approved for shipment at Room Temperature or on Dry Ice
Technology Individual chromatography-purified DNA fragments
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.