Learn More
Invitrogen™ Platinum™ SuperFi™ PCR Master Mix
Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification.
£286.00 - £5380.00
Specifications
GC-Rich PCR Performance | High |
---|---|
Polymerase | Platinum SuperFi DNA Polymerase |
Reaction Speed | Fast |
Shipping Condition | Wet or Dry Ice |
For Use With (Application) | Hot-start PCR, High-fidelity PCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
15525043
|
Invitrogen™
12358010 |
100 Reactions |
£286.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15535043
|
Invitrogen™
12358050 |
500 Reactions |
£1272.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15545043
|
Invitrogen™
12358250 |
5 x 500 Reactions |
£5380.00
2500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Invitrogen Platinum SuperFi PCR Master Mix is a ready-to-use mixture of DNA polymerase, salts, magnesium, and dNTPs for efficient PCR amplification. Only template, primers, and water need to be added, thus reducing the number of pipetting steps during PCR reaction setup.
Platinum SuperFi PCR Master Mix retains all the features of Platinum SuperFi DNA Polymerase, a proofreading DNA polymerase that combines superior fidelity with trusted Platinum hot-start technology. Platinum SuperFi DNA Polymerase is ideally suited for cloning, mutagenesis, and other applications benefiting from supreme sequence accuracy.
Platinum SuperFi DNA Polymerase is engineered with a DNA-binding domain resulting in high processivity and increased resistance to PCR inhibitors. This feature also enables fast-cycling protocols and amplification of long targets. The Platinum hot-start technology is based on proprietary antibodies that inhibit enzyme activity until the initial PCR denaturation step, preventing non-specific amplification and primer degradation.
Features
- Exceptional >300X Taq fidelity
- High specificity and increased yields with Platinum hot-start technology
- Robust amplification of difficult targets of high GC content
- Room temperature reaction setup and 24-hour bench top stability of pre-assembled reactions
- Amplification of samples with suboptimal purity
Applications
- High-fidelity PCR
- Cloning and sub-cloning
- Site-directed mutagenesis
- Amplification of GC-rich templates
- Template generation for sequencing
- High-throughput PCR
- Long PCR
Notes
- Annealing rules for Platinum SuperFi DNA Polymerase are different from many common DNA polymerases.
- Platinum SuperFi PCR Master Mix is supplied with separate SuperFi GC Enhancer for difficult and GC-rich templates (˃65% GC).
Specifications
High | |
Fast | |
Hot-start PCR, High-fidelity PCR | |
Built-In Hot Start | |
SuperMix or Master Mix |
Platinum SuperFi DNA Polymerase | |
Wet or Dry Ice | |
300X | |
Blunt | |
20 kb or less |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.