Learn More
Invitrogen™ 5' RACE System for Rapid Amplification of cDNA Ends, version 2.0
Suitable for rapid amplification of cDNA ends (RACE) between defined point in mRNA and the 5' end
Brand: Invitrogen™ 18374058
722.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: The 5' RACE System is supplied with the components listed in Table 1.
Description
The 5' RACE System provides a set of prequalified reagents intended for synthesis of first-strand cDNA, purification of first-strand products, homopolymeric tailing, and preparation of target cDNA for subsequent amplification by PCR. Control RNA, DNA, and primers are provided for monitoring system performance. The system is useful for the amplification of rare messages for which little sequence information may be available, and for capturing the 5' end information of mRNA.
- Uses advanced capabilities of SuperScriptII RNase H- RT for greater first-strand cDNA yields and increased cDNA lengths
- Prepares specific cDNA from up to 1μg of total or poly(A)+ RNA in one reaction
- Offers improved first-strand cDNA purification procedure that yields cDNA suitable for tailing
- Uses recombinant TdT with optimized reaction conditions in PCR-compatible buffer to provide efficient primer annealing site for PCR
Cloning, PCR and Real-Time PCR, Reverse Transcription, cDNA Libraries and Library Construction
Specifications
cDNA library | |
The 5´ RACE System is supplied with the components listed in Table 1. Store the reagents at -20°C and the DNA purification components at +4°C. | |
cDNA Libraries & Library Construction | |
10 Reactions |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.