Learn More
Invitrogen™ 95401.H1POOL (Yeast Deletion Pools)
Allow simultaneous analysis of large numbers of deletion strains through selective growth conditions
Brand: Invitrogen™ 95401.H1POOL
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: Supplied in five tubes, each containing 200μL of pooled deletions
Description
- Identification and quantitation of deletions surviving selection can be determined by hybridizing deletions to microarray containing all barcode oligos
- Barcode oligos are unique to each deletion, allowing rapid, high-throughput identification of deletions
- Each Yeast Deletion Pool contains all deletions within strain, equally represented within pool
Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Yeast Deletion Mapping
Order Info
Shipping condition: Dry ice
Specifications
Agarose | |
5 x 200 μL Tubes |
For Research Use Only. Not for use in diagnostic procedures.