missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AccuPrime™ GC-Rich DNA Polymerase

Product Code. p-4926955
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10515013

Brand: Invitrogen™ 12337016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

AccuPrime GC-Rich DNA Polymerase (100μL, at 2U/μL) is supplied with two 5X reaction buffers and a separate vial (1mL) of 50mM MgSO4 . Both buffers contain dNTPs, MgSO4 , and AccuPrime accessory proteins. Reaction buffer A (1mL) is for genomic targets. Reaction buffer B (1mL) is for use with cDNA, plasmid, or lambda targets. Sufficient reagents are provided for 200 amplification reactions of 25μL each (at 1 unit of enzyme per reaction).

AccuPrime GC-Rich DNA Polymerase is designed to provide high-yield, high-specificity amplification of difficult-to-amplify templates, such as those with >65% GC content.

AccuPrime GC-Rich DNA Polymerase is designed to provide high-yield, high-specificity amplification of difficult-to-amplify templates, such as those with >65% GC content. The kit offers a choice of buffers for amplifying genomic DNA targets (Buffer A) or non-GC-rich cDNA, plasmid, and lambda-based targets (Buffer B).

AccuPrime GC-Rich DNA Polymerase features

  • High yields for targets up to 5 kb in length
  • AccuPrime accessory proteins for improved PCR specificity
  • Sensitivity down to 5 ng of template DNA

Robust and specific amplifications

The AccuPrime buffers contain thermostable proteins that enhance primer-template hybridization during PCR, increasing the specificity of the reaction. The polymerase itself, from the archaebacterium Pyrolobus fumarius, has a five-fold better processivity than Taq DNA polymerase, and remains active even after 4 hours at 95°C.

Unit definition

One unit of AccuPrime GC-Rich DNA Polymerase is the amount of enzyme required to incorporate 10 nmoles of dNTPs into acid-insoluble material in 30 min. at 74°C.

Order Info

Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • AccuPrime GC-Rich DNA Polymerase (1 x 100 μL at 2 U/μL)
• 5X AccuPrime GC-Rich Buffer A (1 x 1 mL)
• 5X AccuPrime GC-Rich Buffer B (1 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at –20°C.

GC-Rich PCR Performance High
Polymerase AccuPrime GC-Rich DNA Polymerase
Reaction Speed Standard
Quantity 200 rxns
Shipping Condition Wet or Dry Ice
For Use With (Application) Hot-start PCR
Fidelity (vs. Taq) 2X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang Blunt
Reaction Format Separate Components
Size (Final Product) 5 kb or less
Volume 100 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.