missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ AccuPrime™ Taq DNA Polymerase, High Fidelity

Product Code. p-4922147
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10125142

Brand: Invitrogen™ 12346086

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

AccuPrime™ Taq DNA Polymerase High Fidelity (40μL, at 5U/μL)) is supplied with: 50mM MgSO4 (1mL), 10X AccuPrime™ PCR Buffer I (1mL), 10X AccuPrime™ PCR Buffer II (1mL)

Buffer I and II differ in their concentration of thermostable AccuPrime™ protein. Their components are: 600mM Tris-SO4 (pH 8.9), 180mM (NH4)2SO4, 20mM MgSO4, 2mM dGTP, 2mM dATP, 2mM dTTP, 2mM dCTP, thermostable AccuPrime™ protein, 10% glycerol. The supplied PCR Buffer is a 10X concentrate and should be diluted for use. Buffer I is optimized for plasmids, cDNA, and λDNA. Buffer II is optimized for genomic DNA.

Invitrogen AccuPrime Taq DNA Polymerase, High Fidelity, provides qualified reagents for the high-fidelity PCR amplification of nucleic acid templates.

Invitrogen AccuPrime Taq DNA Polymerase, High Fidelity, provides qualified reagents for the high-fidelity PCR amplification of nucleic acid templates. It includes an enzyme blend of Platinum Taq DNA Polymerase and a proofreading enzyme, plus AccuPrime accessory proteins for improved PCR fidelity, yield, and specificity over other hot-start DNA polymerases.

Features of AccuPrime Taq DNA Polymerase, High Fidelity

  • High specificity and yield for robust PCR amplification
  • 9-fold higher fidelity than Taq DNA polymerase alone
  • Mimimal optimization steps, even with non-optimized primer sets
  • Efficient amplification of targets over a broad size range up to 20 kb

How it works

Pyrococcus species GB-D polymerase is a proofreading enzyme that possesses a 3' to 5' exonuclease activity. Mixing this enzyme with Taq DNA Polymerase increases fidelity and allows amplification of simple and complex DNA templates over a large range of target sizes.

The Platinum antibody complexes with Taq DNA polymerase and inhibits activity at room temperature. Activity is restored after the initial denaturation step, providing an automatic hot start PCR.

The thermostable AccuPrime accessory proteins enhance specific primer-template hybridization during every cycle of PCR, preventing mispriming and enhancing PCR specificity and yield.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • AccuPrime Taq DNA Polymerase, High Fidelity (1 x 40 μL at 5 U/μL)
• 10X AccuPrime PCR Buffer I (1 x 1 mL)
• 10X AccuPrime PCR Buffer II (1 x 1 mL)
• 50 mM MgSO4 (1 x 1 mL)

Store at -10°C to -30°C.
Detection Method Primer-probe
GC-Rich PCR Performance Low
Polymerase AccuPrime Taq DNA Polymerase High Fidelity
Reaction Speed Standard
Product Type DNA Polymerase
Quantity 200 rxns
Shipping Condition Dry Ice
For Use With (Application) Hot-start PCR, High-fidelity PCR
Fidelity (vs. Taq) 9X
Hot Start Built-In Hot Start
No. of Reactions 200 Reactions
Overhang Mixed
Reaction Format Separate Components
Size (Final Product) 20 kb or less
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.