Learn More
Invitrogen™ Bio-11-UTP (10 mM)
Ambion Bio-11-UTP 10 mM (Biotinylated UTP) is intended for use as substrate in in vitro transcription reactions.
Brand: Invitrogen™ AM8450
160.50 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
Ambion Bio-11-UTP 10 mM (Biotinylated UTP) is intended for use as substrate in in vitro transcription reactions. These modified nucleotides can be utilized by a variety of RNA polymerases, including T7, T3, and SP6 RNA polymerases, as well as other RNA modifying enzymes. It is provided in one tube containing 25 μL (250 nmol).
Applications
Biotinylated RNA can be used in place of radioactively labeled RNA in many applications, including cRNA amplification and labeling for microarray target preparation. Detection is then carried out via one of a variety of streptavidin-based methods.
Order Info
Shipping Conditions: Dry Ice
Specifications
Nucleotides | |
Dry Ice | |
Biotin | |
Ambion™ | |
Liquid |
Bio-11-UTP, 10 mM (25 μL) Store at –70°C. |
|
10 mM | |
25 μL | |
Nucleic Acid Labeling, Oligo Synthesis |
For Research Use Only. Not for use in diagnostic procedures.