missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bio-16-UTP (10 mM)

Ambion Bio-16-UTP 10 mM (Biotinylated UTP) is intended for use as substrate in in vitro transcription reactions.

Brand:  Invitrogen™ AM8452

229.34 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


258.65 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10126864

  • £344.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Ambion Bio-16-UTP 10 mM (Biotinylated UTP) is intended for use as substrate in in vitro transcription reactions. These modified nucleotides can be utilized by a variety of RNA polymerases, including T7, T3, and SP6 RNA polymerases, as well as other RNA modifying enzymes. It is provided in one tube containing 25 μL (250 nmol).

Applications
Biotinylated RNA can be used in place of radioactively labeled RNA in many applications, including cRNA amplification and labeling for microarray target preparation. Detection is then carried out via one of a variety of streptavidin-based methods.

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Nucleotides
RNA Labeling
10 mM
25 μL
Nucleic Acid Labeling, Oligo Synthesis
Bio-16-UTP, 10 mM (25 μL)

Store at –70°C.
Dry Ice
Biotin
Ambion™
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Bio-16-UTP (10 mM) > 25uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.