missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oligo (dT) Primer (50 μM)

Product Code. 10249034
Click to view available options
Quantity:
80 μL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10249034

Brand: Invitrogen™ AM5730G

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

These are the same Invitrogen primers that are currently included in the SuperScript IV Kit (Cat. No. 18091050).

These are the same Invitrogen primers that are currently included in the SuperScript IV Kit (Cat. No. 18091050). They are provided at a stock concentration of 50 μM.

Order Info

Shipping Conditions: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Product Type Primer
Content And Storage Store at –20°C.
Shipping Condition Dry Ice
Concentration 50 μM
Primer Random
Quantity 80 μL
Product Line Ambion
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.