missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNA Century™ Marker Templates

Contain mixtures of linearized plasmids ready for use as templates as part of in vitro transcription reactions for synthesis of labeled RNA molecular size standards

Brand:  Invitrogen™ AM7780

Product Code. 10669075

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

When transcribed with T7 RNA polymerase, the templates produce five RNA transcripts with the lengths of 100, 200, 300, 400, and 500 bases. They can be labeled with radioisotopes, biotin, or other nonisotopic modified nucleotides. They are ideal as size standards for ribonuclease protection assays and other applications where a low molecular weight RNA ladder is useful.

Acrylamide Gel Electrophoresis, DNA & RNA Purification & Analysis, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting

Order Info

Shipping Conditions: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNA Marker
Denaturing Polyacrylamide Gels
100 to 500 bp
5 μg
Nucleic Acid Gel Electrophoresis & Blotting
Store at –20°C.
Room Temperature
Unconjugated
Ambion™
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.