missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ siPORT™ NeoFX™ Transfection Agent with manual
Lipid-based formulation can be used to efficiently transfect adherent cells with siRNA as they are subcultured—without increased cytotoxicity
Marque: Invitrogen™ AM4510M
200.67 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Includes: One tube containing 0.4mL
Description
- Versatile-works with a broad range of cell lines
- Reproducible-produces consistent results, lot-to-lot, plate-to-plate, and well-to-well
- Efficient-performs with high siRNA transfection efficiency, allowing use of low siRNA concentrations to minimize nonspecific effects
- Fast-reverse-transfects cells as they are plated, saving a day
- Reverse-Transfect Cells as They are Plated, Saving a Day
- Optimized for Lower siRNA Concentrations and High Reproducibility
- Compatible with a Broad Range of Cell Lines
Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection
Informations sur la commande
Shipping Condition: Room Temperature
Spécification
Store at 4°C. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
High-throughput Compatible | |
Transfection Reagent | |
Yes |
Stem Cells, Primary Cells, Mammalian | |
Synthetic siRNA | |
Transfection | |
Ambion™, siPORT™ | |
400 μL |
For Research Use Only. Not for use in diagnostic procedures.