missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ siPORT™ NeoFX™ Transfection Agent with manual
Lipid-based formulation can be used to efficiently transfect adherent cells with siRNA as they are subcultured—without increased cytotoxicity
£290.00 - £532.00
Specifications
Content And Storage | Store at 4°C. |
---|---|
Cell Type | Stem Cells, Primary Cells, Mammalian |
Format | 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks |
Sample Type | Synthetic siRNA |
Transfection Technique | Lipid-based Transfection |
Includes: One tube containing 0.4mL
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
10218704
|
Invitrogen™
AM4510 |
Transfection Reagent | 400 μL |
£290.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10197234
|
Invitrogen™
AM4510M |
Transfection Reagent | 400 μL |
£301.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137004
|
Invitrogen™
AM4511 |
Transfection Reagent | 1 mL |
£532.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005124
|
Invitrogen™
AM4511M |
Transfection Reagent | 1 mL | ||||||
Description
- Versatile-works with a broad range of cell lines
- Reproducible-produces consistent results, lot-to-lot, plate-to-plate, and well-to-well
- Efficient-performs with high siRNA transfection efficiency, allowing use of low siRNA concentrations to minimize nonspecific effects
- Fast-reverse-transfects cells as they are plated, saving a day
- Reverse-Transfect Cells as They are Plated, Saving a Day
- Optimized for Lower siRNA Concentrations and High Reproducibility
- Compatible with a Broad Range of Cell Lines
Cell Culture, RNAi Transfection, Synthetic siRNA Transfection, Transfection
Order Info
Shipping Condition: Room Temperature
Specifications
Store at 4°C. | |
6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks | |
Lipid-based Transfection | |
High-throughput Compatible | |
Transfection Reagent |
Stem Cells, Primary Cells, Mammalian | |
Synthetic siRNA | |
Transfection | |
Ambion™, siPORT™ | |
Yes |
For Research Use Only. Not for use in diagnostic procedures.