missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bac-to-Bac™ Vector Kit

Designed for fast, small-scale production of recombinant baculovirus

Brand:  Invitrogen™ 10360014

Product Code. 10022712

  • £814.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: 10μg of pFastBac1 vector and pFastBac 1-Gus control vector.
This item is not returnable. View return policy

Description

Description

  • Contains pFastBac™1 vector, as well expression control vector, intended for use as part of Bac-to-Bac Baculovirus Expression System, which enables efficent production of recombinant baculovirus for expression testing in insect cells
  • Relies on generation of recombinant baculovirus by site-specific transposition in E. colirather than homologous recombination in insect cells
  • pFastBac 1 vector provided by this kit offers strong polyhedrin promoter for protein expression and large multiple cloning site for simplified cloning
  • Time-saving expression bacmid: expression cassette of pFastBac vector recombines with parent bacmid in DH10BacE. coli
  • Competent Cells to form expression bacmid: bacmid is then transfected into insect cells for production of recombinant baculovirus particles
  • Easy colony screening
  • Parent bacmid in DH10Bac E. colicontains segment of lacZα gene
  • lacZα gene is disrupted upon transposition of expression cassette into bacmid allowing for blue/white selection of recombinants

Baculovirus Production and Titering, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyhedrin
Contains 10 μg of pFastBac™1 vector and pFastBac™ 1-Gus control vector.

Store at -20°C.
Restriction Enzyme/MCS
pFastBac
Vector Kit
Untagged
1 kit
Bac-to-Bac™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Bac-to-Bac™ Vector Kit > 1 kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.