Learn More
Invitrogen™ BenchPro™ 4100 Western Card
Disposable, single-use card that is inserted into the slot on top of the BenchPro™ 4100 Card Processing Station
Brand:  Invitrogen™ WP1001
Product Code. 10455225
  • £140.00 / Pack of 10
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Includes: Box of 10 Cards and 10 membrane holders
Description
Description
Developed for immunodetection, the card holds the membrane to be processed in a Blot Holder. During operation, reagents are pumped from reagent bottles and vials located in the reagent tray, into the card and around the membrane. The order of reagents used and timing of protocol steps are based on the protocol selected.
Compatible for use with WP0001.
Order Info
Shipping Condition: Room Temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Box of 10 Cards and 10 membrane holders. Store at room temperature.
Western Blot
BenchPro
Card
10 cards
1 Membrane/Card
BenchPro™ 4100 Western Processing System, XCell SureLock™ Mini, iBlot™
1 Membrane/Card
Compatible Compatible Products WP0001
Room Temperature