Learn More
Invitrogen™ BioPrime™ Total Genomic Labeling Module
Supports a complete genomic DNA labeling system for use in array comparative genomic hybridization applications (aCGH) that allow for better call rates with precious samples
Brand: Invitrogen™ 18097012
Code : 50
Additional Details : Weight : 0.45000kg
Description
The BioPrime Total Genomic Labeling Module is part of the system that offers users the following advantages:
- Higher signal to noise with Alexa Fluor™ 3 and 5 dyes
- Less channel bias due to novel reaction formulation
- Widest range of input material (50ng - 3μg)
- Simplified workflow with master mix formulation
Contents and Storage:
The BioPrime™ Total Genomic Labeling Module contains Alexa Fluor 3 and Alexa Fluor 5 2x reaction mix, Exo-Klenow fragment (40 U/μL), 5mM EDTA, TE Buffer, and control DNA (Salmon Sperm). Available in a direct labeling format.
Array CGH, ChIP-on-Chip, Chromatin Biology, DNA and RNA Purification and Analysis, DNA Labeling, Gene Expression Analysis and Genotyping, Genotyping and Genomic Profiling, Nucleic Acid Labeling and Oligo Synthesis, RNAi, Epigenetics and Non-Coding RNA Research
Specifications
Fluorescence | |
Probes (Labeled DNA) | |
Yes | |
BioPrime™ | |
30 Reactions |
Alexa Fluor 3, Alexa Fluor 5 | |
Genomic DNA | |
Direct Labeling | |
Total Genomic Labeling Module |
For Research Use Only. Not for use in diagnostic procedures.