missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-14-dATP

Product Code. 10484552
Click to view available options
Quantity:
50 nmol
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10484552

Brand: Invitrogen™ 19524016

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Biotin-14-dATP is supplied at 0.4mM in 100mM Tris-HCl (pH 7.5), 0.1mM EDTA.

A patented dATP analog with biotin attached at the 6-position of the purine base by a 14-atom linker

The long, 14-atom spacer arm allows accessibility to streptavidin for sensitive detection of probe-target hybrids in in situ and filter-based hybridizations. Biotin-14-dATP can be efficiently incorporated into DNA by nick translation in the presence of dCTP, dGTP, and dTTP. Biotin-14-dATP can be added to the 3' end of DNA using terminal deoxynucleotidyl transferase. The amount supplied is sufficient for labeling up to 50μg of DNA by nick translation.

Performance and Quality Testing: Purity is evaluated by reverse-phase HPLC analysis.

DNA and RNA Purification and Analysis, DNA Labeling, Nucleic Acid Labeling and Oligo Synthesis

Order Info

Shipping Condition: Approved for shipment on Wet or Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.4 mM
Content And Storage Biotin-14-dATP is supplied at 0.4 mM in 100 mM Tris-HCl (pH 7.5), 0.1 mM EDTA. Store at -20°C.
Format Liquid
Labeling Method Direct Labeling
Label or Dye Biotin and Other Hapten
Product Type Labeled Nucleotide
Quantity 50 nmol
Shipping Condition Approved for shipment on Wet or Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.