missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ One Shot™ BL21-AI™ Chemically Competent E. coli

Product Code. 11540146
Click to view available options
Quantity:
21 x 50 μL/tube
Unit Size:
1000µL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11540146

Brand: Invitrogen™ C607003

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

BL21-AI Chemically Competent E. coli are provided in single-use, 50-μL aliquots, with S.O.C. medium and supercoiled control plasmid. Store competent cells at -80°C. All components are guaranteed stable for 6 months when properly stored.

One Shot BL21-AI Chemically Competent E. coli cells are designed for applications that require tight regulation and strong expression of toxic proteins from any T7-promoter-based expression systems.

One Shot BL21-AI Chemically Competent E. coli cells are designed for applications that require tight regulation and strong expression of toxic proteins from any T7-promoter-based expression systems. A derivative of the E. coli B strain, BL21-AI does not contain the lon protease and is deliberately deficient in the outer membrane protease OmpT.

The lack of two key proteases reduces degradation of heterologous proteins expressed in the strain. Additionally, several other strain features make these cells especially well suited for protein production, namely fast growth in minimal medium, a lower acetate production when grown in high glucose media, and the ability to reach high cell density.

BL21-AI E. coli contain a chromosomal insertion of a cassette containing the T7 RNA polymerase (T7RNAP) into the araB locus of the araBAD operon, placing regulation of T7 RNAP under the control of the arabinose-inducible araBAD promoter. The araB gene is deleted in this strain. The presence of the tetA gene confers resistance to tetracycline and permits verification of strain identity using tetracycline. The yield of recombinant protein obtained from BL21-AI is generally similar to that obtained from BL21 (DE3). BL21-AI, due it’s qualities and genomic background, was used to elucidate fundamental steps in the adaptation process of the CRISPR/Cas system. One Shot BL21-AI Chemically Competent cells have a transformation efficiency of >1 x 108 cfu/μg plasmid DNA.

One Shot BL21-AI Chemically Competent E. coli offer:
• Transformation efficiency of >1 x 108 cfu/μg control plasmid DNA
• Ion and OmpT protease deficiency that reduces degradation of recombinant protein
• T7 RNA polymerase controlled by the tightly regulated arabinose-inducible araBAD promoter
hsdS mutation that allows efficient transformation of unmethylated DNA
• Ideal for inducing expression of toxic protein from any T7 promoter-based plasmid

Easy-to-use One Shot format
BL21-AI Chemically Competent E. coli cells are supplied in the convenient, single-reaction One Shot format. The single-tube, single-use format allows all steps of the transformation protocol, up to plating, to take place in the same tube, thereby helping save time and prevent contamination.

Genotype
FompT hsdSB (rB, mB) gal dcm araB::T7RNAP-tetA

Find the strain and format that fits your needs
We offer several E. coli strains for protein expression.
BL21 Star (DE3) and other strains are available in MultiShot formats for high throughput applications.
We offer a variety of systems for the expression of recombinant proteins in E. coli. The Champion pET expression system provides the highest level of protein production available in any expression system.

Notes
• To induce expression from the araBAD promoter, use L-arabinose. To repress basal expression from the araBAD promoter, use glucose. Even if glucose is absent, basal expression from is generally low.
• BL21-AI strain genome sequences available in the NCBI GenBank database under accession number CP047231 (Bhawsinghka et al. 2020).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemically Competent Cells
Contains F' Episome No
Improves Plasmid Quality No
Cloning Methylated DNA No
Transformation Efficiency Level Medium Efficiency (1 x 108 to 1 x 109 cfu/μg)
Content And Storage • BL21-AI One Shot Chemically Competent E. coli (21 x 50 μL); store at –80°C
• 20% L-arabinose in sterile water; store at –80°C
• pUC19 Control DNA (50 μL); store at –80°C
• S.O.C. Medium (6 mL); store at 4°C or room temperature

Antibiotic Resistance Bacterial Yes (Tetracycline)
Blue/White Screening No
High-throughput Compatibility Low
Preparing Unmethylated DNA Yes (dcm)
Reduces Recombination No
Shipping Condition Dry Ice
T1 Phage - Resistant (tonA) No
Species E. coli (B)
Promoter T7
Format Tube
Product Line One Shot
Quantity 21 x 50 μL/tube
Toxic Proteins Yes (araB)
Improves RNA Stability No
Improves Protein Stability Yes (lon, ompT)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.