Learn More
Invitrogen™ BloodPrep™ DNA Purification Solution
The BloodPrep™ DNA Purification Solution is a cell lysis and DNA purification solution for whole blood from any animal species, cultured cells, and buccal swabs for use with BloodPrep™ DNA chemistry.
Brand: Invitrogen™ 4342775
Additional Details : Weight : 0.00450kg
Description
The BloodPrep™ DNA Purification Solution is a cell lysis and DNA purification solution for whole blood from any animal species, cultured cells, and buccal swabs for use with BloodPrep™ DNA chemistry.
For Research Use Only. Not for use in diagnostics procedures.
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
Nucleic Acid Purification | |
BloodPrep™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.