missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bovine Serum Albumin

Product Code. 11558896
Click to view available options
Quantity:
150 mg
Unit Size:
150mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11558896

Brand: Invitrogen™ 15561020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Suitable for use as enzyme stabilizer

Bovine Serum Albumin is suitable for use as enzyme stabilizer during purification or for dilution of restriction endonucleases and nucleic acid modifying enzymes. BSA is also commonly used in DNA and protein labeling experiments as a blocking agent to minimize background.

  • Acetylated to inactivate contaminating nucleases and proteases
  • Supplied as aqueous solution at concentration of 45 to 55mg/mL
  • Performance and quality testin: Purity is determined by SDS-PAGE
  • No detectable contaminating activity is observed in 3' and 5' exonuclease, DNA nicking, and restriction endonuclease inhibition assays

ChIP-on-Chip, Chromatin Biology, Cloning, RNAi, Epigenetics & Noncoding RNA Research, Restriction Enzyme Cloning

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 45 to 55 mg/mL
Content And Storage Bovine Serum Albumin is supplied as an aqueous solution at a concentration of 45 to 55 mg/ml. Store at -20°C. Bovine Serum Albumin is guaranteed for 6 months from date of purchase unless otherwise specified in product literature.
Protein BSA
Shipping Condition Approved for shipment on Wet or Dry Ice
Quantity 150 mg
Product Type Bovine Serum Albumin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.