Learn More
Invitrogen™ Champion™ pET302/NT-His and pET303/CT-His Vector Kit
Designed for cloning gene of interest via restriction enzymes and ligase (REaL) and subsequent high-level expression from strong bacteriophage T7 promoter
Brand: Invitrogen™ K630203
Code : 50
Additional Details : Weight : 0.02000kg
Description
- In addition to T7 promoter, each vector contains only necessary functional elements and N- or C-terminal 6xHis tag (pET302/NT-His and pET303/CT-His, respectively) for convenient purification and detection
Bacterial Expression, Bioproduction, Protein Expression, Proteins, Expression, Isolation and Analysis
Specifications
Inducible | |
T7 | |
None | |
His Tag (6x) | |
pET, pDEST |
IPTG | |
DNA Cloning Vector Kit | |
Ampicillin (AmpR) | |
Restriction Enzyme/MCS | |
Champion™ |
For Research Use Only. Not for use in diagnostic procedures.