missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™ Forensic DNA Purification Kit

Product Code. 10614123
Click to view available options
No. of Reactions:
100 Preps
960 Preps
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10614123

Brand: Invitrogen™ CS1120010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

ChargeSwitch Lysis Buffer (L13)
ChargeSwitch Magnetic Beads
Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
ChargeSwitch Purification Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

ChargeSwitch™ Forensic DNA Purification Kit uses magnetic bead technology to allow for rapid and efficient purification of genomic DNA from a variety of forensic samples, including poor or environmentally degraded samples, and is fully validated for Single Tandem Repeat (STR) analysis.

ChargeSwitch™ Forensic DNA Purification Kit uses magnetic bead technology to allow for rapid and efficient purification of genomic DNA from a variety of forensic samples, including poor or environmentally degraded samples, and is fully validated for Single Tandem Repeat (STR) analysis.

• Optimized for higher-purity DNA extraction
• Validated for STR profiling on diverse forensic samples-set
• Compatible with high-throughput protocols

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

All components of the ChargeSwitch™ Forensic DNA Purification Kits are shipped at room temperature. Upon receipt, store the Proteinase K at 4°C. Store all other components at room temperature. All components are guaranteed stable for 6 months, if stored properly.

Components:
• ChargeSwitch™ Lysis Buffer (L13)
• ChargeSwitch™ Magnetic Beads
• Proteinase K (20 mg/mL in 50 mM Tris-HCl, pH 8.5, 5 mM CaCl2, 50% glycerol)
• ChargeSwitch™ Purification Buffer (N5)
• ChargeSwitch™ Wash Buffer (W12)
• ChargeSwitch™ Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)

Format Kit
Isolation Technology ChargeSwitch Chemistry
Sample Type Forensic Sample
For Use With (Application) Forensic
High-throughput Compatibility High-throughput Compatible
No. of Reactions 960 Preps
Product Line ChargeSwitch
Product Type Forensic DNA Purification Kit
Quantity 960 Preps
Shipping Condition Room Temperature
Target Genomic DNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.