Learn More
Invitrogen™ ChargeSwitch™ PCR Clean-Up Kit
ChargeSwitch PCR Clean-Up Kit is the fastest and most effective method of removing primers, unincorporated dNTPs, thermostable polymerases and salts.
£204.00 - £1358.00
Specifications
Content And Storage | The ChargeSwitch™ PCR Clean-Up Kit is shipped at room temperature. Upon receipt, store all components at room temperature. All components are guaranteed stable for 6 months when stored properly. |
---|---|
Format | Kit |
High-throughput Compatibility | High-throughput Compatible |
Product Line | ChargeSwitch™ |
Product Type | PCR Clean-Up Kit |
Includes: ChargeSwitch Magnetic Beads (25 mg/mL in 10 mM MES, pH 5.0, 10 mM NaCl, 0.1% Tween 20)
ChargeSwitch Purification Buffer (N5)
ChargeSwitch Wash Buffer (W12)
ChargeSwitch Elution Buffer (E5; 10 mM Tris-HCl, pH 8.5)
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
10685273
|
Invitrogen™
CS12000 |
100 Preps |
£204.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10221853
|
Invitrogen™
CS1200010 |
960 Preps |
£1358.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ChargeSwitch PCR Clean-Up Kit is the fastest and most effective method of removing primers, unincorporated dNTPs, thermostable polymerases and salts (Figure 1). This kit is scalable for use on liquid-handling robots. This kit includes all the necessary reagents for the number of preps listed and requires the use of a magnetic accessory. Please see the Accessories Chapter (Chapter 6) to select the best magnetic accessory for your needs.
• Sample size: 25–50 μL
• High recovery of DNA fragments
• Rapid, one tube, 5 minute protocol
Order Info
Shipping Condition: Room Temperature
Specifications
The ChargeSwitch™ PCR Clean-Up Kit is shipped at room temperature. Upon receipt, store all components at room temperature. All components are guaranteed stable for 6 months when stored properly. |
|
High-throughput Compatible | |
PCR Clean-Up Kit | |
Room Temperature | |
Next-Generation Sequencing (NGS), Cloning, Standard PCR | |
PCR Products |
Kit | |
ChargeSwitch™ | |
5 min | |
DNA from Enzymatic Reactions (e.g. PCR) | |
ChargeSwitch™ Chemistry |
For Research Use Only. Not for use in diagnostic procedures.