Learn More
Invitrogen™ ChargeSwitch™-Pro Plasmid Miniprep Kit
The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.
Brand: Invitrogen™ CS30250
Includes: Miniprep Columns with Collection Tubes, Recovery Tubes, Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer
Description
The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.
• 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
• Cleaner purified product for improved performance in downstream applications
• Ready-to-use, complete kit for a fast, hassle-free start
• Ideal for purifying plasmid DNA from 1-5 mL overnight bacterial cultures, yielding up to 20 μg of plasmid DNA
Order Info
Shipping Condition: Room Temperature
Specifications
Spin Column | |
Kit | |
Bacterial Culture | |
Next-Generation Sequencing, Transfection, Cloning, Sequencing, Transformation, Nucleic Acid Labeling, PCR, In Vitro Transcription | |
250 Preps | |
ChargeSwitch™ | |
250 Preps | |
Plasmid DNA |
Store at room temperature. | |
ChargeSwitch Chemistry | |
Plasmid DNA | |
Not High-throughput Compatible (Manual) | |
< 100 μg (Small-Scale) Plasmid DNA | |
Plasmid MiniPrep Kit | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.