missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ChargeSwitch™-Pro Plasmid Miniprep Kit

The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.

Brand:  Invitrogen™ CS30010

 View more versions of this product

Product Code. 10797483

  • £26.33 / Each

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers

Includes: Miniprep Columns with Collection Tubes, Recovery Tubes, Resuspension Buffer, RNase A, Lysis Buffer, Precipitation Buffer, Wash Buffers 1 and 2, and Elution Buffer
This item is not returnable. View return policy

Description

Description

The ChargeSwitch™-Pro Plasmid Miniprep Kit generates high-quality plasmid DNA without the use of harsh or potentially interfering reagents, for better success in downstream applications.

• 100% ethanol-free, guanidinium-free, organics-free, aqueous protocol for a safer, more streamlined method
• Cleaner purified product for improved performance in downstream applications
• Ready-to-use, complete kit for a fast, hassle-free start
• Ideal for purifying plasmid DNA from 1-5 mL overnight bacterial cultures, yielding up to 20 μg of plasmid DNA

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Spin Column
Kit
Mini
Plasmid DNA
Not High-throughput Compatible (Manual)
< 100 μg (Small-Scale) Plasmid DNA
Plasmid MiniPrep Kit
Room Temperature
Store at room temperature.
ChargeSwitch Chemistry
Bacterial Culture
PCR, Southern Blotting, Sequencing, Cloning
10 Preps
ChargeSwitch™
10 Preps
Plasmid DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.