missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ BP Clonase™ Enzyme Mix
£770.00 - £3260.00
Specifications
Shipping Condition | Dry Ice |
---|---|
Enzyme | BP Clonase |
Compatible Buffer | Reaction Buffer |
Product Type | BP Clonase Enzyme Mix |
Product Line | Clonase™, Gateway™ |
Includes: Proteinase K solution (2μg/μL), 30% PEG 8000/30mM MgCl2 Solution and a positive control vector.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10207432
|
Invitrogen™
11789013 |
20 Reactions |
£770.00
20 reactions |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10003152
|
Invitrogen™
11789021 |
100 Reactions |
£3260.00
100 reactions |
Estimated Shipment: 31-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Proteins catalyze in vitro recombination of PCR products or DNA segments from clones (containing attB sites) and Donor vector (containing attP sites) to generate Entry clones
- BP Clonase II enzyme format is single mix of both enzyme and buffer to ensure enzyme stability and enable convenient 10μL reaction set up with fewer pipetting steps, which both maximize recombination efficiency and minimize pippetting
- Original format of BP Clonase enzyme is stand alone enzyme blend and reaction buffer provided in separate tubes
Cloning, Gateway Cloning, Gateway Recombination
Order Info
Shipping Conditions: Dry ice
Specifications
Dry Ice | |
Reaction Buffer | |
Clonase™, Gateway™ |
BP Clonase | |
BP Clonase Enzyme Mix |
For Research Use Only. Not for use in diagnostic procedures.