missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gateway™ BP Clonase™ II Enzyme mix
Click to view available options
Quantity:
100 Reactions
20 Reactions
Packungsgröße:
100 reactions
20 reactions
Beschreibung
Beinhaltet
Proteinase K solution (2μg/μL) and a positive control vector
- Catalyzes the in vitro recombination of PCR products or subcloning DNA segments from clones (containing attB sites) and a donor vector (containing attP sites) to generate entry clones
- Contains enzymes and buffer in a single mix to enable convenient ten-microliter reaction set up with fewer pipetting steps
- Guaranteed stable for 6 months when properly stored at -20°C to -80°C
Bestellinformation
Shipping Conditions: Dry ice
Spezifikation
Spezifikation
| Content And Storage | Gateway™ BP Clonase™ II enzyme mix includes proteinase K solution (2 μg/μl) and a positive control vector. Store at -20°C or -80°C. Guaranteed stable for 6 months when properly stored. |
| Shipping Condition | Dry Ice |
| Enzyme | BP Clonase |
| Compatible Buffer | Enzyme Buffer |
| Quantity | 100 Reactions |
| Product Type | BP Clonase Enzyme Mix |
| Product Line | Clonase, Gateway |
For Research Use Only. Not for use in diagnostic procedures.
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur