missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CloneChecker™ System (for screening bacterial colonies)

CloneChecker System is a rapid method for screening recombinant bacterial colonies or liquid cultures of colonies for the presence of target plasmid DNA.

Brand:  Invitrogen™ 11666013

79.80 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10155262

  • £140.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 

Includes: The CloneChecker System includes Yellow, Red, and Green solutions and Loading buffer.
This item is not returnable. View return policy

Description

Description

CloneChecker System is a rapid method for screening recombinant bacterial colonies or liquid cultures of colonies for the presence of target plasmid DNA.

  • Screens bacterial colonies for plasmid DNA and insert of interest prior to purification
  • Screens for inserts >200 bp in plasmids up to 15 kb by size comparison to the original vector or a DNA size marker
  • Allows for supercoiled, restriction digest, or PCR analysis
  • Works with single-copy plasmids starting with a 2mm bacterial colony
  • Includes sufficient reagents for processing 100 bacterial samples by supercoiled analysis (direct size comparison); and 100 samples by restriction endonuclease or PCR analysis 

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

The CloneChecker™ System includes Yellow, Red, and Green solutions and Loading buffer.

Store at 15° to 30°C.
Plasmid DNA
Not High-throughput Compatible (Manual)
CloneChecker™
100 preps
Plasmid DNA
Bacterial Culture
Cloning, PCR
< 100 μg (Small-Scale) Plasmid DNA
Bacterial Colony Screening System
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.