Learn More
Invitrogen™ Countess™ Automated Cell Counter, with 101 Boxes of Slides
Benchtop instrument for easy cell and viability counts in as little as 30 seconds
Brand:  Invitrogen™ C10227
 View more versions of this product
Product Code. 10389053
  • £5070.00 / Each
Explore more special offers
For Research Use Only. All usage must comply with product instructions.
Includes:
  • One Countess Automated Cell Counter, 101 boxes of 50 slides each (1050 total slides), 2 x 100mL bottles of Trypan blue stain, Power cord with 4 adaptors and Quick Reference Guide (C10311)
  • One Countess Automated Cell Counter, one box of 50 slides (100 total slides), 2 x 1mL vials of Trypan blue stain, power cord with 4 adaptors, and Quick Reference Guide (C10227)
  • One Countess Automated Cell Counter, 11 boxes of 50 slides each (550 total slides), 22 x 1mL vials of Trypan blue stain, Power cord with 4 adaptor and Quick Reference Guide (C10310)
Description
Description
  • For verifying Number of Cells, Live Cells/mL, Dead Cells/mL, Cell Size
  • Compatible with Countess cell counting chamber slides
  • Dimensions: D x W x H: 20 x 27 x 19cm (7.8 x 10.6 x 7.4 in.)
Cell Counting, Viability and Cryopreservation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 x 27 x 19cm
One Countess Automated Cell Counter, one box of 50 slides (100 total slides), 2 x 1mL vials of Trypan blue stain, power cord with 4 adaptors, and Quick Reference Guide
100-240 VAC
50/60 Hz
Automated Cell Counter with 1 box of slides
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products