missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ CD34 Positive Isolation Kit

Use this kit to isolate a high purity and yield of human CD34+ progenitor stem cells.

Brand:  Invitrogen™ 11301D

652.80 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10796073

  • £768.00 / 5mL
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: Each kit contains 5mL Dynabeads coated with anti-CD34 monoclonal antibody, and 5mL DETACHaBEAD™ CD34. The kit can process up to 5x109 cells
This item is not returnable. View return policy

Description

Description

Use this kit to isolate a high purity and yield of human CD34+ progenitor stem cells. Positively isolated cells are bead and antibody-free, phenotypically unaltered and suitable for any downstream applications including flow cytometry, functional studies, and cell culture to produce dendritic cells (DC's). The Dynal™ CD34 Progenitor Cell Isolation System can also be used to isolate CD34+ cells from rhesus monkey (Macaca mulatta).

• Positive isolation of human CD34+ progenitor stem cells with bead release
• Stem cells can be isolated directly from whole blood, cord blood or bone marrow
• Isolated CD34+ cells can be used in any application, i.e. be derived into dendritic cells (DC's)

Starting samples:
PBMC from mobilized peripheral blood, bone marrow or cord blood.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Stem Cells, Stem Cells (Hematopoietic)
Positive Cell Isolation Kit
Processes ∼5 x 109 cells total
DETACHaBEAD™, DYNAL™, Dynabeads™
5 mL
Room Temperature
Human
Each kit contains 5 mL Dynabeads™ coated with anti-CD34 monoclonal antibody, and 5 mL DETACHaBEAD™ CD34.

The kit can process up to 5 x 109 cells. Store components at 2°C to 8°C.
Positive Isolation
>95%
Research Grade
Blood (Mobilized Peripheral), Blood (Umbilical Cord), Bone Marrow
1 x 108 PBMCs per isolation
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.