Learn More
Invitrogen™ MAX Efficiency™ DH10Bac Competent Cells
Capable of supporting site-specific recombination between pFastBac™ and bMON14272
Brand: Invitrogen™ 10361012
351.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.05000kg
Includes: MAX Efficiency DH10Bac Chemically Competent Cells: 5 vials, 100μL each (total of 500μL), pUC19 DNA (0.01 μg/mL): 1 vial, 100μL, SOC Medium: 2 bottles, 6mL each
Description
MAX Efficiency DH10Bac Competent Cells are used for production of recombinant bacmids used in the Bac-to-Bac™ Baculovirus Expression System. The DH10Bac E. coli strain contains a baculovirus shuttle vector (bacmid) that can recombine with a donor plasmid, pFastBac™, to create an expression bacmid containing a cloned gene of interest.
Using MAX Efficiency DH10Bac competent cells
MAX Efficiency DH10Bac Competent Cells can serve as a host for a recombinant pFastBac™ vector containing a cloned gene of interest. DH10Bac cells harbor a baculovirus shuttle vector (bMON14272) and a helper plasmid (pMON7142), and can generate high molecular weight bacmids that can then be amplified, purified, and used for insect cell transfection and subsequent gene expression. Kanamycin resistance for bacmid selection and maintenance is conferred by bMON14272, and tetracycline resistance by pMON7124
- Consistent yields of >1 x 108 transformants/μg
- Tolerance of small amounts of undiluted ligation reactions
- Support of efficient blue/white colony screening (Φ80lacZΔM15 marker)
Not High Throughput-Compatible (Manual)
Baculovirus Production and Titering, Cloning, Insect Expression, Protein Expression, Proteins, Expression, Isolation and Analysis, Transformation
Order Info
Shipping Condition: Dry Ice
Specifications
Competent Cells | |
> 1x10^8 | |
No | |
Medium Efficiency (10^8-10^9 cfu⁄μg) | |
Not Suitable for cloning unstable DNA | |
Yes | |
Not Suitable for preparing unmethylated DNA | |
No | |
No | |
Tube | |
5 x 100 μL |
Lacks F' Episome | |
Chemically Competent | |
No | |
Contains: • MAX Efficiency DH10Bac Chemically Competent Cells: 5 vials, 100 μl each (total of 500 μl) • pUC19 DNA (0.01 μg/ml): 1 vial, 100 μl • SOC Medium: 2 bottles, 6 ml each Store Competent Cells at -80°C. Store pUC19 DNA at -20°C. Store SOC Medium at 4°C or at room temperature. |
|
DH10Bac | |
Not High-throughput Compatible (Manual) | |
Not for ccdB vector propagation | |
Dry Ice | |
E. coli | |
DH10Bac, MAX Efficiency™ |
For Research Use Only. Not for use in diagnostic procedures.