Learn More
Invitrogen™ λ DNA (cIind 1 ts857 Sam 7)
λ DNA (cIind 1 ts857 Sam 7) is isolated from a dam+ dcm+ E coli LE597 lysogen
Brand: Invitrogen™ 25250010
207.91 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01000kg
Includes: Supplied at 0.35 to 0.6μg/μL in 10mM Tris-HCl (pH 7.4), 5mM NaCl, 0.1mM EDTA.
Description
Suitable for use as a substrate for generating molecular size standards and for testing restriction endonuclease activity. Restriction endonucleases sensitive to dam or dcm methylation will not cleave this DNA.
Performance and Quality Testing: Purity, DNA structure, and selected restriction endonuclease fragmentation patterns are verified by agarose gel analysis.
Agarose Gel Electrophoresis, Cloning, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting, Restriction Enzyme Cloning
Order Info
Shipping Condition: Wet Ice
Specifications
λ DNA | |
Supplied at 0.25 to 0.6 μg/μl in 10 mM Tris-HCl (pH 7.4), 5 mM NaCl, 0.1 mM EDTA. Store at +4°C. | |
Wet Ice | |
500 μg |
Control Template(s) | |
Unconjugated | |
Tube |
For Research Use Only. Not for use in diagnostic procedures.