missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DNA Polymerase I

Product Code. p-4921658
Click to view available options
Quantity:
1000 U
250 U
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10358002

Brand: Invitrogen™ 18010017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

1000 units of DNA polymerase in a storage buffer of 50mM potassium phosphate (pH 7.0), 100mM KCl, 1mM DTT, and 50% (v/v) glycerol. It is supplied at 5-10 units/μL.

DNA polymerase with 5'→3' and 3'→5' exodeoxyribonuclease activities

Performance and quality testing
Endodeoxyribonuclease assay; efficiency of DNase I-dependent nick translation determined.

Unit definition

One unit incorporates 10nMol of total deoxyribonucleotide into acid-precipitable material in 30 min at 37°C using template/primer.

Unit reaction conditions

50mM potassium phosphate (pH 7.0), 6.7mM MgCl2 , 1mM 2-mercaptoethanol, 80μg/mL template/primer, 32μM dTTP, 69nM [3H]dTTP, and enzyme in 100μL for 30 min at 37°C. Also incorporates biotinylated nucleotides.

  • Applications: DNase I-dependent nick translation, second-strand synthesis in cDNA cloning, fill-in of 5' overhangs
  • Source: purified from E. coli expressing the DNA Polymerase I gene on a plasmid

Cloning, Restriction Enzyme Cloning

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -30°C).
Description DNA Polymerase
Form Liquid
Polymerase DNA Polymerase I
Exonuclease Activity 3' - 5', 5' - 3'
Quantity 250 U
Shipping Condition Wet Ice
Hot Start No

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.