missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Exosome-Human CD81 Isolation Reagent (from cell culture)

Obtain a highly pure exosome subset with easy protocol scalability with Exosome-Human CD81 Isolation Reagent (from cell culture).

Brand:  Invitrogen™ 10616D

358.70 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15460544

  • £422.00 / 2mL
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Obtain a highly pure exosome subset with easy protocol scalability with Exosome-Human CD81 Isolation Reagent (from cell culture). Following isolation, the specific sub-population of exosomes, ECVs can be further studied using methods such as Western blotting, qPCR, or next generation sequencing.

  • Minimal hands-on time for specific exosome isolation
  • Compatible with most downstream analysis
  • Recent studies suggest that exosome/ECV population consists of several subsets, possibly with different functions
  • Using immuno-affinity based Dynabeads magnetic separation technology, these subsets can be selectively isolated and studied
  • Cell culture media must first be enriched for exosomes
  • Can be done quickly and efficiently using Total Exosome Isolation Reagent (from cell culture media) or traditional ultracentrifugation
  • Following enrichment, specific exosomes are isolated by binding to surface of magnetic beads
  • In addition to their high-quality sensitivity, reproducibility, and stability, Dynabeads allows you to see samples during handling due to color of magnetic beads
  • When a sample tube is placed in the magnet, bead-bound exosomes are pulled to side of tube, allowing for easy separation and buffer changes
  • Sample and bead volume can be easily scaled according to your sample size or downstream applications

Mix thoroughly to ensure that magnetic beads do not settle in the sample tube. A mixer which provides both tilting and rotation for optimal mixing is recommended.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C).
Dynabeads™
Cell Cultures
Isolation Reagent
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.