missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ Untouched™ Human T Cells Kit

Product Code. 10033093
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10033093

Brand: Invitrogen™ 11344D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

10mL Depletion Dynabeads and 2mL Antibody Mix with monoclonal antibodies towards human CD14, CD16 (a and b), CD19, CD36, CD56, CD123 and CD235a (Glycophorin A)

Use this kit to isolate pure and viable untouched T-cells from PBMC by negative isolation. The kit depletes B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes.

Use this kit to isolate pure and viable untouched T-cells from PBMC by negative isolation. The kit depletes B cells, NK cells, monocytes, platelets, dendritic cells, granulocytes and erythrocytes. The negatively isolated human T-cells are left in the sample and have not been in contact with the Dynabeads. The bead and antibody-free T-cells are truly untouched, and in perfect shape for any functional assay and application.

• High purity, recovery and viability of human untouched T cells
• Easy to use and to scale up
• No columns required

Methodology:
An antibody mix towards the non-T cells is added to the sample and allowed to bind to the cells. Dynabeads™ are added and will bind to the antibody-labeled cells during a short incubation. The bead-bound cells are quickly separated on a magnet and discarded. The remaining negatively isolated and untouched human T-cells can be directly analyzed in a flow cytometer and used in any application.

Note: A negative isolation strategy isolates pure and truly untouched cells. When maximum cell purity is critical, a positive cell isolation strategy is recommended

Starting Sample:
PBMC

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Cell Type T Cells (Whole Population)
Content And Storage The Kit Contains: 10 mL Depletion Dynabeads™ and 2 mL Antibody Mix with monoclonal antibodies towards human CD14, CD16 (a and b), CD19, CD36, CD56, CD123 and CD235a (Glycophorin A).

Store at 2°C to 8°C.
Product Type Cell Isolation Kit
Final Product Type Cells
Isolation Technology Negative Isolation
No. of Cells Processes ∼1 x 109 cells total
No. of Tests 100 Isolations
Output Viability >95%
Product Line DYNAL, Dynabeads, Untouched
Quantity 1 kit
Sample Type PBMC
Shipping Condition Room Temperature
Starting Material Cell No. 1 x 107 PBMCs per isolation
Target Species Human
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.