missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ MyOne™ Streptavidin T1

Dynabeads MyOne Streptavidin T1 beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for use in the isolation and handling of biotinylated nucleic acids, antibodies, and other biotinylated ligands.

Brand:  Invitrogen™ 65602

1313.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15544364

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Supplied in PBS, pH 7.4 with 0.1% BSA and 0.02% NaN3 added as preservatives.
This item is not returnable. View return policy

Description

Description

Dynabeads MyOne Streptavidin T1 beads are 1-μm monosized, monodispersed, superparamagnetic beads covalently coupled to streptavidin and intended for use in the isolation and handling of biotinylated nucleic acids, antibodies, and other biotinylated ligands. Their small but uniform size presents a high surface area per mg of beads and a correspondingly high capacity for the target ligand, while the high iron content ensures an efficient magnetic pull. These features, along with a slow sedimentation rate during incubation, contribute to a gentle and rapid magnetic separation. Excellent dispersion abilities and the lack of magnetic remanence make Dynabeads streptavidin products ideal for manual and automated protocols, including microfluidic system protocols.

Benefits of Dynabeads MyOne Streptavidin T1 beads include:
Performance—high sensitivity and reproducibility secure consistent results
Efficiency—fast binding kinetics and high binding capacity
Flexibility—flexible protocols with gentle and efficient liquid-phase reaction kinetics
Automation ready—easy automation for high-throughput needs on the Kingfisher Purification System

Consistency and reproducibility
The monolayer of streptavidin leaves most biotin-binding sites available for the binding of biotin and biotinylated ligands. The specific and defined surface allows for efficient capture, separation, and downstream handling. In addition, the low size distribution (<5%) of the monosized beads and the streptavidin monolayer ensure negligible leakage, providing batch consistency and reproducibility of results.

High binding capacity
The size of the molecule and the biotinylation procedure will affect the binding capacity. The capacity also depends on steric availability and charge interaction between bead and molecule and between molecules. With two or three biotin binding sites available for each streptavidin molecule on the surface of the bead after immobilization, one mg of Dynabeads MyOne Streptavidin T1 typically binds:
• 950–1,500 pmol free biotin 
• ∼20 μg biotinylated IgG 
• ∼400 pmol biotinylated peptides 
• ∼20 μg biotinylated dsDNA 
• ∼400 pmol biotinylated ss-oligonucleotides

Applications
Thermo Fisher streptavidin-coupled Dynabeads magnetic beads have been used and cited for a wide variety of applications. Although the beads can be used to isolate any biotinylated targets, these hydrophobic beads are particularly well suited for immunoassays, protein purification, cell isolation, phage display, and biopanning.

Automation ready with the KingFisher Purification System 
The 1-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher Sample Purification Systems or liquid handlers.

Commercial supply 
Our manufacturing sites are ISO 13485–certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility making them an ideal choice for commercial supply. If you are in the process of customizing Dynabeads MyOne Streptavidin T1 beads in a commercial product or service or need a larger bulk volume of this product, please contact us atoemdynal@lifetech.com or see our Dynabeads OEM page.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Magnetic Polystyrene
10 mg/mL
Recombinant streptavidin covalently coupled to Dynabeads
Magnetic Beads
Any Sample Type
Capture of Biotinylated Targets
Dynabeads™, MyOne™
10 mL
Recombinant
950–1,500 pmoles free biotin/mg beads
ISO9001 and ISO13485
Tosylactivated, Hydrophobic
Brown
Dynabeads MyOne Streptavidin T1 are supplied in PBS, pH 7.4, with 0.1% BSA and 0.02% NaN3 . Store at 2–8°C.

Beads in suspension
1 μm
KingFisher™ Sample Purification System, DynaMag™ magnets
Yes
Streptavidin Magnetic Beads
Ambient Temperature
Biotin
For Research Use Only
Streptavidin
CV <5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.