Learn More
Invitrogen™ E-Gel™ NGS™ 0.8% Agarose Gels
Designed to provide fast, convenient, and easy electrophoresis
Brand: Invitrogen™ A25798
133.00 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
E-Gel NGS 0.8% Agarose Gels are self-contained, pre-cast agarose gels designed to provide fast, convenient, and easy electrophoresis. Each E-Gel agarose gel is ready for use with agarose, electrodes, and SYBR Safe DNA gel stain packaged inside an 11-well, easy-to-open, UV- and blue-light transparent cassette. SYBR Safe stain can be viewed with blue light, which minimizes DNA damage and allows safe, real-time monitoring of DNA migration using the E-Gel Power Snap Electrophoresis System.
Easy access to the gel
The gel cassette is designed to open with a gel knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis.
Running and viewing apparatus
E-Gel NGS gels require the E-Gel Power Snap Electrophoresis System for running and viewing the gel in real time.
Specifications
E-Gel™ 1 Kb Plus DNA Ladder | |
0.8% | |
Agarose Gel | |
SYBR™ Safe | |
Less hazardous |
Store at room temperature. | |
E-Gel | |
800 to 10,000 bp | |
11-well | |
Single Comb |
For Research Use Only. Not for use in diagnostic procedures.