missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ EX Agarose Gels, 2%

Precast, fast-resolving gels with 11 wells and an openable format

Brand:  Invitrogen™ G402002

Product Code. 10053894

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Contain a proprietary fluorescent nucleic acid stain (excitation: 490nm, emission: 520nm) that can be viewed with blue light, which minimizes DNA damage and allows detection down to 1ng/band of DNA
  • Complete separation (by molecular weight) in 10 minutes
  • Live monitoring of DNA migration without the hazards of UV light
  • Easy access to the gel for extraction of DNA bands or transfer to membranes for Southern blotting
  • Demonstrate over five-fold greater sensitivity than comparable gels containing ethidium bromide allows less sample use per run
  • Fast analysis of RNA samples to check on their integrity before proceeding with downstream application
  • The gel cassette is designed to open with a Gel Knife, so that the gel inside can be accessed readily for excision of specific bands or transferred to a membrane for Southern blot analysis
  • Single comb well format
  • Sample Type: Double-Stranded DNA (dsDNA), Single-Stranded RNA
  • Sample Volume; 20μL

Agarose Gel Electrophoresis, DNA and RNA Purification and Analysis, Nucleic Acid Gel Electrophoresis and Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

E-Gel™ EX 20-Paks contain 20 gels. Store gels at room temperature. E-Gel™ EX gels are guaranteed stable for 3 months when properly stored.
E-Gel
20 μL
11-well
E-Gel EX 20-Paks contain 20 gels.
E-Gel™
Preparative without dyes, Non-preparative
Room Temperature
2%
EX Agarose Gel
100 to 2000 bp
Proprietary Stain
Molecular Weight
20 Pack
Double-Stranded DNA (dsDNA), Single-Stranded RNA
Single Comb
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers